Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291501.1 | NP_001278430.1 | 23 | 3464 | ||
NM_148674.4 | NP_683515.4 | 25 | 3686 | |||
XM_011530144.2 | XP_011528446.1 | 25 | 3547 | |||
GenBank mRNA | AJ504806.2 | - | 25 | 3686 | ||
AK125736.1 | - | 12 | 1445 | |||
BC126208.1 | - | 23 | 3444 | 8991924 |