Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015697.7 | NP_056512.5 | 7 | 1171 | ||
XM_017008031.1 | XP_016863520.1 | 7 | 1294 | |||
GenBank mRNA | AF091086.1 | - | 7 | 839 | ||
AJ621061.3 | - | 7 | 1171 | |||
AK129583.1 | - | 7 | 927 | |||
BC008804.2 | - | 7 | 1028 | 3954787 | ||
BC020728.1 | - | 7 | 1068 | 4696019 | ||
BC116454.1 | - | 2 | 215 | |||
BQ363759.1 | - | 4 | 447 |