Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292002.1 | NP_001278931.1 | 4 | 420 | ||
NM_014487.5 | NP_055302.1 | 5 | 491 | |||
XM_017008033.1 | XP_016863522.1 | 5 | 635 | |||
GenBank mRNA | AJ006591.2 | - | 5 | 436 | ||
AK002106.1 | - | 5 | 441 | |||
AK223609.1 | - | 5 | 442 | |||
AK294089.1 | - | 4 | 374 | |||
AK315463.1 | - | 5 | 477 | |||
BC004421.1 | - | 5 | 467 | 3352916 | ||
DA089047.1 | - | 5 | 483 |