Search
Search

NKIRAS1| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020345.3 | NP_065078.1 | 4 | 722 | ||
| XM_005265076.3 | XP_005265133.1 | 4 | 609 | |||
| XM_005265077.4 | XP_005265134.1 | 3 | 577 | |||
| XM_005265078.3 | XP_005265135.1 | 4 | 561 | |||
| GenBank mRNA | AF229839.1 | - | 2 | 320 | ||
| AK027749.1 | - | 4 | 699 | |||
| BC012145.1 | - | 4 | 605 | 4549097 | ||
| BC066940.1 | - | 4 | 613 | 5273965 | ||