Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256182.1 | NP_001243111.1 | 10 | 1750 | ||
NM_001256183.1 | NP_001243112.1 | 9 | 1676 | |||
NM_013275.5 | NP_037407.4 | 9 | 1679 | |||
XM_006721181.1 | XP_006721244.1 | 9 | 1456 | |||
XM_006721184.2 | XP_006721247.1 | 8 | 1494 | |||
XM_011523051.2 | XP_011521353.1 | 10 | 1739 | |||
XM_011523053.2 | XP_011521355.1 | 10 | 1845 | |||
XM_011523054.2 | XP_011521356.1 | 10 | 1759 | |||
XM_011523055.2 | XP_011521357.1 | 11 | 2044 | |||
XM_011523056.2 | XP_011521358.1 | 10 | 1781 | |||
XM_011523057.2 | XP_011521359.1 | 9 | 1662 | |||
XM_017023182.1 | XP_016878671.1 | 9 | 2112 | |||
XM_017023183.1 | XP_016878672.1 | 10 | 1848 | |||
XM_017023184.1 | XP_016878673.1 | 10 | 1845 | |||
XM_017023185.1 | XP_016878674.1 | 8 | 1486 | |||
XM_017023186.1 | XP_016878675.1 | 12 | 2222 | |||
XM_017023187.1 | XP_016878676.1 | 8 | 1574 | |||
GenBank mRNA | AB209186.1 | - | 8 | 1323 | ||
AF121775.1 | - | 9 | 1435 | |||
AK290994.1 | - | 8 | 1358 | |||
AY131227.1 | - | 1 | 75 | |||
AY373756.1 | - | 9 | 1682 | |||
AY533563.1 | - | 10 | 1507 | |||
BC017437.1 | - | 9 | 1648 | |||
BC052290.1 | - | 9 | 1594 | |||
BC069013.1 | - | 9 | 1594 |