Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RACGAP1 SIRNA Assays ›
Gene Symbol:
RACGAP1
Gene Name:
Rac GTPase activating protein 1
Gene Aliases:
CYK4, HsCYK-4, ID-GAP, MgcRacGAP
Chromosome Location:
Chr.12: 49989162 - 50033136 on Build GRCh38
UniGene ID:
Hs.505469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26551
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RACGAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001126103.2
NP_001119575.1
6
708
NM_001126104.2
NP_001119576.1
4
588
NM_001319999.1
NP_001306928.1
4
518
NM_001320000.1
NP_001306929.1
5
592
NM_001320001.1
NP_001306930.1
4
584
NM_001320002.1
NP_001306931.1
4
543
NM_001320003.1
NP_001306932.1
4
514
NM_001320004.1
NP_001306933.1
4
465
NM_001320005.1
NP_001306934.1
3
429
NM_001320006.1
NP_001306935.1
5
588
NM_001320007.1
NP_001306936.1
3
315
NM_013277.4
NP_037409.2
6
697
XM_006719359.1
XP_006719422.1
4
416
XM_011538238.1
XP_011536540.1
4
373
XM_011538243.1
XP_011536545.1
4
493
XM_017019220.1
XP_016874709.1
4
690
XM_017019221.1
XP_016874710.1
3
334
XM_017019222.1
XP_016874711.1
3
289
XM_017019223.1
XP_016874712.1
3
330
XM_017019224.1
XP_016874713.1
3
260
XM_017019225.1
XP_016874714.1
3
169
XM_017019226.1
XP_016874715.1
3
229
XM_017019227.1
XP_016874716.1
3
250
GenBank mRNA
AB030251.1
-
4
424
AB040911.1
-
2
169
AK000733.1
-
4
431
AK225901.1
-
3
359
AK315785.1
-
6
645
AL136794.1
-
6
578
BC024144.2
-
4
409
BC032754.1
-
4
415
5583315
BU940828.1
-
5
482
CR533565.1
-
3
354
DA051287.1
-
4
422
DA441098.1
-
4
486
DA500396.1
-
3
235
DA502516.1
-
4
400
DA812983.1
-
5
496
DB076571.1
-
4
485
DB084617.1
-
4
466
DB090068.1
-
3
367
DB164810.1
-
4
401
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
actomyosin contractile ring assembly
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
spermatogenesis
neuroblast proliferation
sulfate transport
embryo development
antigen processing and presentation of exogenous peptide antigen via MHC class II
positive regulation of cytokinesis
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
mitotic spindle midzone assembly
regulation of attachment of spindle microtubules to kinetochore
GTPase activator activity
protein binding
phosphatidylinositol-3,4,5-trisphosphate binding
microtubule binding
protein kinase binding
alpha-tubulin binding
gamma-tubulin binding
metal ion binding
beta-tubulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0