Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BLNK SIRNA Assays ›
          Gene Symbol:
          BLNK
          Gene Name:
          B-cell linker
          Gene Aliases:
          AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
          Chromosome Location:
          Chr.10: 96189171 - 96271576 on Build GRCh38
          UniGene ID:
          Hs.665244
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s26556

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BLNK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114094.1 NP_001107566.1 4, 5 393
          NM_001258440.1 NP_001245369.1 4, 5 333
          NM_001258441.1 NP_001245370.1 4, 5 333
          NM_001258442.1 NP_001245371.1 4, 5 333
          NM_013314.3 NP_037446.1 4, 5 393
          NR_047680.1 4, 5 333
          NR_047681.1 4, 5 338
          NR_047682.1 4, 5 333
          NR_047683.1 4, 5 333
          XM_011539728.2 XP_011538030.1 4, 5 270
          XM_017016159.1 XP_016871648.1 4, 5 271
          GenBank mRNA AF068180.1 - 4, 5 368
          AF068181.1 - 4, 5 368
          AK027779.1 - 4, 5 328
          AK225546.1 - 4, 5 376
          AM180327.1 - 4, 5 215
          AM180328.1 - 4, 5 215
          AM180329.1 - 4, 5 215
          AM180330.1 - 4, 5 215
          AM180331.1 - 4, 5 215
          AM180332.1 - 4, 5 215
          AM180333.1 - 4, 5 215
          AM180334.1 - 4, 5 215
          AM180336.1 - 4, 5 215
          AM180337.1 - 4, 5 215
          AM180338.1 - 4, 5 220
          AM180339.1 - 4, 5 215
          AM180340.1 - 4, 5 215
          AM180341.1 - 3, 4 165
          AM180342.1 - 3, 4 165
          AM180343.1 - 3, 4 165
          AM180344.1 - 2, 3 124
          AM180345.1 - 3, 4 165
          AM180346.1 - 2, 3 163
          BC018906.2 - 4, 5 307 4053894
          BC110298.1 - 4, 5 325
          BM820310.1 - 4, 5 386
          BU597768.1 - 4, 5 338
          DA045252.1 - 4, 5 393

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001025767, BC098790.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          humoral immune response
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of signal transduction
          B cell differentiation
          intracellular signal transduction
          transmembrane receptor protein tyrosine kinase adaptor activity
          SH3/SH2 adaptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline