Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SENP1 SIRNA Assays ›
          Gene Symbol:
          SENP1
          Gene Name:
          SUMO1/sentrin specific peptidase 1
          Gene Aliases:
          SuPr-2
          Chromosome Location:
          Chr.12: 48042893 - 48106308 on Build GRCh38
          UniGene ID:
          Hs.371957
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s26615

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SENP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001267594.1 NP_001254523.1 8 1132
          NM_001267595.1 NP_001254524.1 8 1387
          NR_051991.1 9 1573
          NR_051992.1 9 1586
          XM_006719361.3 XP_006719424.1 8 1267
          XM_006719362.2 XP_006719425.1 9 1039
          XM_011538244.2 XP_011536546.1 8 1307
          XM_011538245.2 XP_011536547.1 7 1219
          XM_017019229.1 XP_016874718.1 9 1405
          XM_017019230.1 XP_016874719.1 9 1360
          XM_017019231.1 XP_016874720.1 9 1305
          XM_017019232.1 XP_016874721.1 9 1032
          XM_017019233.1 XP_016874722.1 8 1315
          XM_017019234.1 XP_016874723.1 8 1225
          XM_017019235.1 XP_016874724.1 8 958
          XM_017019236.1 XP_016874725.1 8 1232
          XM_017019237.1 XP_016874726.1 8 1132
          XM_017019238.1 XP_016874727.1 9 1343
          XM_017019239.1 XP_016874728.1 8 1383
          XM_017019240.1 XP_016874729.1 9 1343
          GenBank mRNA AF149770.1 - 6 807
          AK292060.1 - 8 903
          AK312874.1 - 8 1132
          BC045639.1 - 8 1203 5298667
          BX537920.1 - 9 1586
          BX640784.1 - 9 1417

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of definitive erythrocyte differentiation
          protein sumoylation
          protein desumoylation
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of transcription from RNA polymerase II promoter
          apoptotic signaling pathway
          endopeptidase activity
          protein binding
          SUMO-specific protease activity
          SUMO-specific endopeptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sxctq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0