Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034837.2 | NP_001030009.1 | 8 | 1142 | ||
NM_001034838.2 | NP_001030010.1 | 7 | 989 | |||
NM_001278339.1 | NP_001265268.1 | 7 | 1184 | |||
NM_001278340.1 | NP_001265269.1 | 8 | 1271 | |||
NM_014592.3 | NP_055407.1 | 7 | 1109 | |||
XM_017009407.1 | XP_016864896.1 | 8 | 1113 | |||
GenBank mRNA | AF199597.1 | - | 7 | 572 | ||
AK074390.1 | - | 7 | 705 | |||
AK301775.1 | - | 7 | 764 | |||
AY170821.1 | - | 7 | 785 | |||
AY780424.1 | - | 8 | 947 | |||
BC035032.1 | - | 7 | 1114 | |||
BC050375.1 | - | 7 | 876 | 5752207 | ||
DQ148476.1 | - | 7 | 599 | |||
DQ148477.1 | - | 7 | 572 | |||
DQ148478.1 | - | 8 | 605 | |||
DQ148479.1 | - | 7 | 647 |