Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014601.3 | NP_055416.2 | 5 | 1266 | ||
GenBank mRNA | AF181263.1 | - | 4 | 1054 | ||
AF454952.1 | - | 4 | 1071 | |||
AK074701.1 | - | 5 | 1188 | |||
AK097126.1 | - | 5 | 593 | |||
AK097228.1 | - | 4 | 585 | |||
AK293429.1 | - | 1 | 115 | |||
AK296907.1 | - | 6 | 1158 | |||
AK298067.1 | - | 4 | 729 | |||
AK300512.1 | - | 2 | 217 | |||
AK315548.1 | - | 5 | 1190 | |||
AK316448.1 | - | 2 | 166 | |||
BC014445.1 | - | 5 | 1177 | 4908085 |