Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD63 SIRNA Assays ›
          Gene Symbol:
          CD63
          Gene Name:
          CD63 molecule
          Gene Aliases:
          LAMP-3, ME491, MLA1, OMA81H, TSPAN30
          Chromosome Location:
          Chr.12: 55725443 - 55729673 on Build GRCh38
          UniGene ID:
          Hs.445570
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2700

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD63
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257389.1 NP_001244318.1 5, 6 824
          NM_001257390.1 NP_001244319.1 5, 6 567
          NM_001257391.1 NP_001244320.1 4, 5 562
          NM_001257392.1 NP_001244321.1 4, 5 363
          NM_001257400.1 NP_001244329.1 4, 5 392
          NM_001257401.1 NP_001244330.1 4, 5 532
          NM_001267698.1 NP_001254627.1 5, 6 614
          NM_001780.5 NP_001771.1 5, 6 618
          GenBank mRNA AF508304.1 - 4, 5 360
          AK311893.1 - 5, 6 489
          AL582859.3 - 4, 5 437
          BC002349.2 - 5, 6 506 2819554
          BC013017.2 - 5, 6 462 3010594
          BG478842.1 - 5, 6 568
          BG766593.1 - 4, 5 391
          BQ434569.1 - 533
          BT007073.1 - 4, 5 421
          BT020137.1 - 4, 5 421
          BT020138.1 - 4, 5 421
          BU167765.1 - 5, 6 625
          BU190521.1 - 4, 5 557
          CN296813.1 - 5, 6 581
          CN484590.1 - 4, 5 432
          CR542096.1 - 4, 5 421
          EB386351.1 - 5, 6 553
          KF998086.1 - 4, 5 421
          M58485.1 - 5, 6 476
          M59907.1 - 5, 6 471
          S93788.1 - 4, 5 445
          X07982.1 - 5, 6 448

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          platelet degranulation
          cell-matrix adhesion
          negative regulation of epithelial cell migration
          protein transport
          cell migration
          epithelial cell differentiation
          cellular protein localization
          endosome to melanosome transport
          positive regulation of cell adhesion
          pigment granule maturation
          regulation of vascular endothelial growth factor signaling pathway
          regulation of rubidium ion transport
          positive regulation of integrin-mediated signaling pathway
          protein binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cqs7j:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0