Search
Search

CD79A| Gene Symbol: CD79A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001783.3 | NP_001774.1 | 5 | 918 | ||
| NM_021601.3 | NP_067612.1 | 5 | 804 | |||
| GenBank mRNA | AF283770.3 | - | 5 | 761 | ||
| AK223371.1 | - | 5 | 916 | |||
| BC113731.1 | - | 5 | 753 | 8322783 | ||
| BC113733.1 | - | 5 | 753 | 8322785 | ||
| M80462.1 | - | 5 | 769 | |||
| M86921.1 | - | 5 | 766 | |||
| S46706.1 | - | 5 | 740 | |||
| S75217.1 | - | 5 | 733 | |||