Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC35C2 SIRNA Assays ›
Gene Symbol:
SLC35C2
Gene Name:
solute carrier family 35 member C2
Gene Aliases:
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location:
Chr.20: 46349528 - 46364461 on Build GRCh38
UniGene ID:
Hs.593344
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27222
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC35C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281457.1
NP_001268386.1
2
436
NM_001281458.1
NP_001268387.1
3
304
NM_001281459.1
NP_001268388.1
2
436
NM_001281460.1
NP_001268389.1
3
768
NM_015945.11
NP_057029.8
2
436
NM_173073.3
NP_775096.1
2
436
NM_173179.3
NP_775271.1
2
631
XM_011528831.1
XP_011527133.1
2
614
XM_011528832.1
XP_011527134.1
2
908
XM_011528833.1
XP_011527135.1
2
432
XM_011528834.1
XP_011527136.1
2
436
XM_011528835.1
XP_011527137.1
2
436
XM_011528836.1
XP_011527138.1
2
588
XM_011528837.1
XP_011527139.1
2
422
XM_011528838.1
XP_011527140.1
1
543
XM_017027856.1
XP_016883345.1
2
1331
XM_017027857.1
XP_016883346.1
2
859
XM_017027858.1
XP_016883347.1
2
769
XM_017027859.1
XP_016883348.1
2
1136
XM_017027860.1
XP_016883349.1
3
576
XM_017027861.1
XP_016883350.1
2
598
XM_017027862.1
XP_016883351.1
2
436
GenBank mRNA
AF132949.1
-
2
373
AF455052.1
-
2
391
AK222928.1
-
2
376
AK300207.1
-
2
390
AK300798.1
-
2
368
AK301022.1
-
3
246
AK315726.1
-
2
598
BC014191.2
-
2
380
4761663
BC021138.1
-
2
574
4870715
BC025277.1
-
2
632
4865888
BI831689.1
-
3
380
BM846744.1
-
2
402
DA820126.1
-
2
436
DB023133.1
-
3
519
DB246755.1
-
2
391
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00210988_m1
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
negative regulation of gene expression
UDP-glucose transport
protein O-linked fucosylation
positive regulation of Notch signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline