Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015916.4 | NP_057000.2 | 4 | 1224 | ||
NR_024552.1 | 4 | 1108 | ||||
NR_046344.1 | 4 | 1121 | ||||
XM_006717880.2 | XP_006717943.1 | 4 | 1100 | |||
XM_006717883.2 | XP_006717946.1 | 3 | 985 | |||
XM_006717884.3 | XP_006717947.1 | 4 | 1731 | |||
XM_011539848.2 | XP_011538150.1 | 4 | 1756 | |||
XM_017016306.1 | XP_016871795.1 | 4 | 1683 | |||
XM_017016307.1 | XP_016871796.1 | 4 | 1590 | |||
XM_017016308.1 | XP_016871797.1 | 4 | 1225 | |||
XM_017016309.1 | XP_016871798.1 | 3 | 972 | |||
GenBank mRNA | AF131810.1 | - | 4 | 969 | ||
AK022195.1 | - | 4 | 1119 | |||
BC000039.2 | - | 4 | 1137 | 3509518 |