Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SSUH2 SIRNA Assays ›
Gene Symbol:
SSUH2
Gene Name:
ssu-2 homolog (C. elegans)
Gene Aliases:
C3orf32, SSU-2, fls485
Chromosome Location:
Chr.3: 8619400 - 8745035 on Build GRCh38
UniGene ID:
Hs.736935
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27336
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SSUH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256748.1
NP_001243677.1
6, 7
557
NM_001256749.1
NP_001243678.1
6, 7
578
NM_015931.2
NP_057015.1
6, 7
724
NR_046358.1
7, 8
805
XM_011533774.1
XP_011532076.1
14, 15
2273
XM_017006510.1
XP_016861999.1
7, 8
5038
XM_017006511.1
XP_016862000.1
13, 14
1961
XM_017006512.1
XP_016862001.1
13, 14
2130
XM_017006513.1
XP_016862002.1
12, 13
1966
XM_017006514.1
XP_016862003.1
12, 13
1872
XM_017006515.1
XP_016862004.1
14, 15
3027
XM_017006516.1
XP_016862005.1
9, 10
1388
XM_017006517.1
XP_016862006.1
9, 10
1341
XM_017006518.1
XP_016862007.1
14, 15
2252
XM_017006519.1
XP_016862008.1
7, 8
4983
XM_017006520.1
XP_016862009.1
7, 8
1334
XM_017006521.1
XP_016862010.1
10, 11
1492
XM_017006522.1
XP_016862011.1
10, 11
1383
XM_017006523.1
XP_016862012.1
8, 9
1086
XM_017006524.1
XP_016862013.1
9, 10
1324
XM_017006525.1
XP_016862014.1
12, 13
1893
XM_017006526.1
XP_016862015.1
7, 8
972
XM_017006527.1
XP_016862016.1
13, 14
2015
XM_017006528.1
XP_016862017.1
7, 8
1011
XM_017006529.1
XP_016862018.1
7, 8
1041
XM_017006530.1
XP_016862019.1
9, 10
1474
XM_017006531.1
XP_016862020.1
8, 9
1396
XM_017006532.1
XP_016862021.1
7, 8
5896
XM_017006533.1
XP_016862022.1
13, 14
2048
XM_017006534.1
XP_016862023.1
13, 14
1994
XM_017006535.1
XP_016862024.1
7, 8
6172
XM_017006536.1
XP_016862025.1
7, 8
6175
XM_017006537.1
XP_016862026.1
7, 8
6175
XM_017006538.1
XP_016862027.1
11, 12
1591
XM_017006539.1
XP_016862028.1
6, 7
703
XM_017006540.1
XP_016862029.1
7, 8
882
XM_017006541.1
XP_016862030.1
4, 5
524
XM_017006542.1
XP_016862031.1
4, 5
502
GenBank mRNA
AB024705.1
-
6, 7
697
AK057972.1
-
7, 8
801
AK093066.1
-
12, 13
1754
AK122938.1
-
11, 12
1539
AK225925.1
-
7, 8
801
AK300175.1
-
6, 7
557
BC052614.1
-
6, 7
561
6300057
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-p8qt9:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0