Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SSUH2 SIRNA Assays ›
Gene Symbol:
SSUH2
Gene Name:
ssu-2 homolog (C. elegans)
Gene Aliases:
C3orf32, SSU-2, fls485
Chromosome Location:
Chr.3: 8619400 - 8745035 on Build GRCh38
UniGene ID:
Hs.736935
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27337
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SSUH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256748.1
NP_001243677.1
5, 6
438
NM_001256749.1
NP_001243678.1
5, 6
459
NM_015931.2
NP_057015.1
5, 6
605
NR_046358.1
6, 7
686
XM_011533774.1
XP_011532076.1
13, 14
2154
XM_017006510.1
XP_016861999.1
6, 7
4919
XM_017006511.1
XP_016862000.1
12, 13
1842
XM_017006512.1
XP_016862001.1
12, 13
2011
XM_017006513.1
XP_016862002.1
11, 12
1847
XM_017006514.1
XP_016862003.1
11, 12
1753
XM_017006515.1
XP_016862004.1
13, 14
2908
XM_017006516.1
XP_016862005.1
8, 9
1269
XM_017006517.1
XP_016862006.1
8, 9
1222
XM_017006518.1
XP_016862007.1
13, 14
2133
XM_017006519.1
XP_016862008.1
6, 7
4864
XM_017006520.1
XP_016862009.1
6, 7
1215
XM_017006521.1
XP_016862010.1
9, 10
1373
XM_017006522.1
XP_016862011.1
9, 10
1264
XM_017006523.1
XP_016862012.1
7, 8
967
XM_017006524.1
XP_016862013.1
8, 9
1205
XM_017006525.1
XP_016862014.1
11, 12
1774
XM_017006526.1
XP_016862015.1
6, 7
853
XM_017006527.1
XP_016862016.1
12, 13
1896
XM_017006528.1
XP_016862017.1
6, 7
892
XM_017006529.1
XP_016862018.1
6, 7
922
XM_017006530.1
XP_016862019.1
8, 9
1355
XM_017006531.1
XP_016862020.1
7, 8
1277
XM_017006532.1
XP_016862021.1
6, 7
5777
XM_017006533.1
XP_016862022.1
12, 13
1929
XM_017006534.1
XP_016862023.1
12, 13
1875
XM_017006535.1
XP_016862024.1
6, 7
6053
XM_017006536.1
XP_016862025.1
6, 7
6056
XM_017006537.1
XP_016862026.1
6, 7
6056
XM_017006538.1
XP_016862027.1
10, 11
1472
XM_017006539.1
XP_016862028.1
5, 6
584
XM_017006540.1
XP_016862029.1
6, 7
763
XM_017006541.1
XP_016862030.1
3, 4
405
XM_017006542.1
XP_016862031.1
3, 4
383
GenBank mRNA
AB024705.1
-
5, 6
578
AK057972.1
-
6, 7
682
AK093066.1
-
11, 12
1635
AK122938.1
-
10, 11
1420
AK225925.1
-
6, 7
682
AK300175.1
-
5, 6
438
BC052614.1
-
5, 6
442
6300057
DB010779.1
-
5, 6
459
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0