Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SSUH2 SIRNA Assays ›
          Gene Symbol:
          SSUH2
          Gene Name:
          ssu-2 homolog (C. elegans)
          Gene Aliases:
          C3orf32, SSU-2, fls485
          Chromosome Location:
          Chr.3: 8619400 - 8745035 on Build GRCh38
          UniGene ID:
          Hs.736935
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s27338

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SSUH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256748.1 NP_001243677.1 12 1062
          NM_001256749.1 NP_001243678.1 12 1083
          NM_015931.2 NP_057015.1 12 1229
          NR_046358.1 13 1310
          XM_011533774.1 XP_011532076.1 20 2778
          XM_017006510.1 XP_016861999.1 13 5543
          XM_017006511.1 XP_016862000.1 19 2466
          XM_017006512.1 XP_016862001.1 19 2635
          XM_017006513.1 XP_016862002.1 18 2471
          XM_017006514.1 XP_016862003.1 18 2377
          XM_017006515.1 XP_016862004.1 20 3532
          XM_017006516.1 XP_016862005.1 15 1893
          XM_017006517.1 XP_016862006.1 15 1846
          XM_017006518.1 XP_016862007.1 20 2757
          XM_017006519.1 XP_016862008.1 13 5488
          XM_017006520.1 XP_016862009.1 13 1839
          XM_017006521.1 XP_016862010.1 16 1997
          XM_017006522.1 XP_016862011.1 16 1888
          XM_017006523.1 XP_016862012.1 14 1591
          XM_017006524.1 XP_016862013.1 15 1829
          XM_017006525.1 XP_016862014.1 18 2398
          XM_017006526.1 XP_016862015.1 13 1477
          XM_017006527.1 XP_016862016.1 19 2520
          XM_017006528.1 XP_016862017.1 13 1516
          XM_017006529.1 XP_016862018.1 13 1546
          XM_017006530.1 XP_016862019.1 15 1979
          XM_017006531.1 XP_016862020.1 14 1901
          XM_017006533.1 XP_016862022.1 19 2553
          XM_017006534.1 XP_016862023.1 19 2499
          XM_017006538.1 XP_016862027.1 17 2096
          XM_017006539.1 XP_016862028.1 12 1208
          XM_017006540.1 XP_016862029.1 13 1387
          XM_017006541.1 XP_016862030.1 10 1029
          XM_017006542.1 XP_016862031.1 10 1007
          GenBank mRNA AB024705.1 - 12 1202
          AK057972.1 - 13 1306
          AK093025.1 - 1 1771
          AK093066.1 - 18 2259
          AK122938.1 - 17 2827
          AK225925.1 - 13 1306
          AK300175.1 - 12 1062
          BC052614.1 - 12 1066 6300057

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0