Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321102.1 | NP_001308031.1 | 9 | 1887 | ||
NM_016030.5 | NP_057114.5 | 9 | 1924 | |||
XM_011510350.2 | XP_011508652.1 | 8 | 2240 | |||
XM_011510355.2 | XP_011508657.1 | 8 | 865 | |||
XM_011510356.1 | XP_011508658.1 | 8 | 757 | |||
XM_017004257.1 | XP_016859746.1 | 9 | 2354 | |||
GenBank mRNA | AF151845.1 | - | 7 | 653 | ||
AK094181.1 | - | 10 | 1892 | |||
AK098327.1 | - | 9 | 1889 | |||
AK125105.1 | - | 4 | 2804 | |||
BC014164.2 | - | 9 | 1801 | 4548681 | ||
BC017475.1 | - | 8 | 922 | |||
BQ954367.1 | - | 4 | 236 |