Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RMDN1 SIRNA Assays ›
Gene Symbol:
RMDN1
Gene Name:
regulator of microtubule dynamics 1
Gene Aliases:
CGI-90, FAM82B, RMD-1, RMD1
Chromosome Location:
Chr.8: 86467116 - 86514348 on Build GRCh38
UniGene ID:
Hs.145386
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27471
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RMDN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286707.1
NP_001273636.1
8
842
NM_001286719.1
NP_001273648.1
8
881
NM_001317807.1
NP_001304736.1
9
750
NM_016033.2
NP_057117.2
9
971
XM_005250944.3
XP_005251001.1
9
877
XM_005250947.3
XP_005251004.1
8
787
XM_005250948.3
XP_005251005.1
9
779
XM_006716573.3
XP_006716636.1
9
966
XM_011517090.2
XP_011515392.1
9
951
XM_011517091.2
XP_011515393.1
9
912
XM_011517092.2
XP_011515394.1
8
861
XM_011517093.2
XP_011515395.1
9
951
XM_011517094.2
XP_011515396.1
9
951
XM_011517095.2
XP_011515397.1
9
951
XM_011517096.2
XP_011515398.1
9
912
XM_011517097.2
XP_011515399.1
8
861
XM_011517098.2
XP_011515400.1
8
822
XM_017013518.1
XP_016869007.1
9
877
XM_017013519.1
XP_016869008.1
9
912
XM_017013520.1
XP_016869009.1
9
877
XM_017013521.1
XP_016869010.1
9
838
XM_017013524.1
XP_016869013.1
9
779
XM_017013525.1
XP_016869014.1
9
779
XM_017013526.1
XP_016869015.1
9
740
XM_017013527.1
XP_016869016.1
8
876
XM_017013528.1
XP_016869017.1
8
684
GenBank mRNA
AF151848.1
-
9
883
AK000672.1
-
9
879
AK297896.1
-
8
743
AK303122.1
-
8
733
AL561197.3
-
9
724
BC008509.1
-
7
535
BC009671.1
-
9
863
3896168
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0