Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IRAK4 SIRNA Assays ›
          Gene Symbol:
          IRAK4
          Gene Name:
          interleukin 1 receptor associated kinase 4
          Gene Aliases:
          IPD1, IRAK-4, NY-REN-64, REN64
          Chromosome Location:
          Chr.12: 43758913 - 43789543 on Build GRCh38
          UniGene ID:
          Hs.138499
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s27527

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IRAK4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114182.2 NP_001107654.1 6, 7 792
          NM_001145256.1 NP_001138728.1 5, 6 646
          NM_001145257.1 NP_001138729.1 4, 5 598
          NM_001145258.1 NP_001138730.1 3, 4 428
          NM_016123.3 NP_057207.2 5, 6 744
          XM_005268943.3 XP_005269000.1 5, 6 849
          XM_005268944.4 XP_005269001.1 5, 6 813
          XM_005268945.4 XP_005269002.1 5, 6 1057
          XM_005268947.4 XP_005269004.1 6, 7 803
          XM_005268948.2 XP_005269005.1 7, 8 843
          XM_005268949.2 XP_005269006.1 4, 5 703
          XM_006719438.3 XP_006719501.1 6, 7 1105
          XM_006719439.2 XP_006719502.1 5, 6 1210
          XM_006719440.2 XP_006719503.1 5, 6 621
          XM_006719441.2 XP_006719504.1 4, 5 506
          XM_006719442.2 XP_006719505.1 5, 6 510
          XM_006719443.3 XP_006719506.1 4, 5 653
          XM_011538431.2 XP_011536733.1 6, 7 861
          XM_011538432.1 XP_011536734.1 6, 7 897
          XM_011538433.2 XP_011536735.1 5, 6 1476
          XM_017019390.1 XP_016874879.1 7, 8 978
          XM_017019391.1 XP_016874880.1 6, 7 943
          GenBank mRNA AF155118.1 - 5, 6 711
          AF445802.1 - 4, 5 662
          AK000528.1 - 5, 6 691
          AK027301.2 - 5, 6 1239
          AK299944.1 - 4, 5 595
          AK304583.1 - 3, 4 431
          AK314288.1 - 5, 6 733
          AY283670.1 - 4, 5 662
          AY283671.1 - 4, 5 660
          AY340962.1 - 5, 6 727
          AY340963.1 - 6, 7 775
          AY340964.1 - 4, 5 581
          AY340965.1 - 5, 6 629
          AY340966.1 - 6, 7 794
          AY340967.1 - 7, 8 842
          AY730682.1 - 4, 5 662
          BC013316.1 - 5, 6 732 4287014
          DA290809.1 - 4, 5 507
          KU178631.1 - 4, 5 662
          KU178632.1 - 3, 4 516

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          toll-like receptor signaling pathway
          neutrophil mediated immunity
          MyD88-dependent toll-like receptor signaling pathway
          JNK cascade
          cytokine-mediated signaling pathway
          toll-like receptor 9 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          positive regulation of smooth muscle cell proliferation
          neutrophil migration
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          interleukin-1 receptor binding
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0