Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IRAK4 SIRNA Assays ›
          Gene Symbol:
          IRAK4
          Gene Name:
          interleukin 1 receptor associated kinase 4
          Gene Aliases:
          IPD1, IRAK-4, NY-REN-64, REN64
          Chromosome Location:
          Chr.12: 43758913 - 43789543 on Build GRCh38
          UniGene ID:
          Hs.138499
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s27529

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IRAK4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114182.2 NP_001107654.1 10, 11 1258
          NM_001145256.1 NP_001138728.1 9, 10 1112
          NM_001145257.1 NP_001138729.1 8, 9 1064
          NM_001145258.1 NP_001138730.1 7, 8 894
          NM_016123.3 NP_057207.2 9, 10 1210
          XM_005268943.3 XP_005269000.1 9, 10 1315
          XM_005268944.4 XP_005269001.1 9, 10 1279
          XM_005268945.4 XP_005269002.1 9, 10 1523
          XM_005268947.4 XP_005269004.1 10, 11 1269
          XM_005268948.2 XP_005269005.1 11, 12 1309
          XM_005268949.2 XP_005269006.1 8, 9 1169
          XM_006719438.3 XP_006719501.1 10, 11 1571
          XM_006719439.2 XP_006719502.1 9, 10 1676
          XM_006719440.2 XP_006719503.1 9, 10 1087
          XM_006719441.2 XP_006719504.1 8, 9 972
          XM_006719442.2 XP_006719505.1 9, 10 976
          XM_006719443.3 XP_006719506.1 8, 9 1119
          XM_011538431.2 XP_011536733.1 10, 11 1327
          XM_011538432.1 XP_011536734.1 10, 11 1363
          XM_011538433.2 XP_011536735.1 9, 10 1942
          XM_017019390.1 XP_016874879.1 11, 12 1444
          XM_017019391.1 XP_016874880.1 10, 11 1409
          GenBank mRNA AF155118.1 - 9, 10 1177
          AF445802.1 - 8, 9 1128
          AK000528.1 - 9, 10 1157
          AK027301.2 - 9, 10 1705
          AK299944.1 - 8, 9 1061
          AK304583.1 - 7, 8 897
          AK314288.1 - 9, 10 1199
          AY283670.1 - 8, 9 1128
          AY283671.1 - 8, 9 1126
          AY340962.1 - 9, 10 1193
          AY340963.1 - 10, 11 1241
          AY340964.1 - 8, 9 1047
          AY340965.1 - 9, 10 1095
          AY340966.1 - 10, 11 1260
          AY340967.1 - 11, 12 1308
          AY730682.1 - 8, 9 1128
          BC013316.1 - 9, 10 1198 4287014
          KU178631.1 - 8, 9 1128
          KU178632.1 - 7, 8 982

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          toll-like receptor signaling pathway
          neutrophil mediated immunity
          MyD88-dependent toll-like receptor signaling pathway
          JNK cascade
          cytokine-mediated signaling pathway
          toll-like receptor 9 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          positive regulation of smooth muscle cell proliferation
          neutrophil migration
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          interleukin-1 receptor binding
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hdb68:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0