Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286760.1 | NP_001273689.1 | 6 | 703 | ||
NM_016174.4 | NP_057258.3 | 6 | 1179 | |||
XM_005252035.1 | XP_005252092.1 | 8 | 2089 | |||
XM_011518762.1 | XP_011517064.1 | 9 | 2246 | |||
XM_011518763.2 | XP_011517065.1 | 6 | 1797 | |||
XM_017014794.1 | XP_016870283.1 | 10 | 1988 | |||
GenBank mRNA | AB040935.1 | - | 7 | 3692 | ||
AF177203.1 | - | 5 | 547 | |||
AK074519.1 | - | 5 | 615 | |||
AK074523.1 | - | 5 | 589 | |||
AK297929.1 | - | 5 | 675 | |||
AK307753.1 | - | 3 | 457 | |||
AK314294.1 | - | 6 | 750 | |||
BC098432.1 | - | 6 | 777 | 6672104 | ||
BC108698.1 | - | 6 | 790 | |||
BC119698.1 | - | 6 | 652 | 40116832 | ||
BC119699.1 | - | 6 | 652 | 40116834 |