Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016216.3 | NP_057300.2 | 8 | 1236 | ||
GenBank mRNA | AF180919.2 | - | 8 | 1190 | ||
AK000116.1 | - | 3 | 713 | |||
AK094147.1 | - | 9 | 1121 | |||
BC009472.1 | - | 8 | 1164 | 3503627 |