Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297748.1 | NP_001284677.1 | 2 | 364 | ||
NM_001297750.1 | NP_001284679.1 | 4 | 561 | |||
NM_001297751.1 | NP_001284680.1 | 3 | 505 | |||
NM_016511.3 | NP_057595.2 | 3 | 463 | |||
XM_011520687.2 | XP_011518989.1 | 3 | 463 | |||
GenBank mRNA | AF200949.1 | - | 3 | 401 | ||
AK075114.1 | - | 3 | 383 | |||
AK091989.1 | - | 2 | 315 | |||
AK291584.1 | - | 3 | 351 | |||
AY358587.1 | - | 3 | 463 | |||
BC039072.1 | - | 3 | 369 | 5178017 | ||
BC067746.1 | - | 3 | 383 | 30337675 | ||
BM920364.1 | - | 403 | ||||
BQ722745.1 | - | 3 | 392 | |||
BX339075.2 | - | 3 | 401 | |||
BX380212.2 | - | 4 | 418 |