Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKMY1 SIRNA Assays ›
Gene Symbol:
ANKMY1
Gene Name:
ankyrin repeat and MYND domain containing 1
Gene Aliases:
ZMYND13
Chromosome Location:
Chr.2: 240468632 - 240561110 on Build GRCh38
UniGene ID:
Hs.656615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27836
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKMY1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282771.1
NP_001269700.1
4
571
NM_001282780.1
NP_001269709.1
4
571
NM_001282781.1
NP_001269710.1
4
571
NM_001308375.1
NP_001295304.1
4
810
NM_016552.3
NP_057636.2
3
408
NM_017844.3
NP_060314.2
4
571
XM_011511300.2
XP_011509602.1
4
755
XM_011511301.1
XP_011509603.1
4
755
XM_011511304.2
XP_011509606.1
4
755
XM_011511305.2
XP_011509607.1
4
755
XM_011511309.2
XP_011509611.1
4
755
XM_011511313.2
XP_011509615.1
4
755
XM_011511315.2
XP_011509617.1
4
755
XM_011511317.1
XP_011509619.1
4
755
XM_011511318.1
XP_011509620.1
4
755
XM_011511319.2
XP_011509621.1
4
755
XM_011511322.1
XP_011509624.1
4
755
XM_017004271.1
XP_016859760.1
4
755
XM_017004272.1
XP_016859761.1
4
755
XM_017004273.1
XP_016859762.1
4
755
XM_017004274.1
XP_016859763.1
4
755
XM_017004275.1
XP_016859764.1
4
521
XM_017004276.1
XP_016859765.1
5
842
XM_017004277.1
XP_016859766.1
4
755
XM_017004278.1
XP_016859767.1
4
755
XM_017004279.1
XP_016859768.1
4
755
XM_017004280.1
XP_016859769.1
4
755
XM_017004281.1
XP_016859770.1
3
361
XM_017004282.1
XP_016859771.1
3
605
XM_017004283.1
XP_016859772.1
5
842
XM_017004284.1
XP_016859773.1
4
755
XM_017004285.1
XP_016859774.1
4
521
XM_017004286.1
XP_016859775.1
3
605
XM_017004287.1
XP_016859776.1
3
605
XM_017004288.1
XP_016859777.1
3
605
XM_017004289.1
XP_016859778.1
3
605
XM_017004290.1
XP_016859779.1
3
605
XM_017004291.1
XP_016859780.1
5
792
GenBank mRNA
AB034636.1
-
3
338
AK000506.1
-
4
538
AK125881.1
-
1
2580
AK225534.1
-
4
538
AK296714.1
-
3
324
AK314534.1
-
4
555
AL136618.1
-
3
317
BC033495.1
-
4
571
BC073146.1
-
4
512
5743806
CR936600.1
-
3
306
CR936759.1
-
4
479
KU178642.1
-
3
413
KU178643.1
-
3
413
Back To Top
More Information
Gene Ontology Categories:
Process(es)
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0