Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKMY1 SIRNA Assays ›
Gene Symbol:
ANKMY1
Gene Name:
ankyrin repeat and MYND domain containing 1
Gene Aliases:
ZMYND13
Chromosome Location:
Chr.2: 240468632 - 240561110 on Build GRCh38
UniGene ID:
Hs.656615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27837
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKMY1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282771.1
NP_001269700.1
3
446
NM_001282780.1
NP_001269709.1
3
446
NM_001282781.1
NP_001269710.1
3
446
NM_001308375.1
NP_001295304.1
3
685
NM_016552.3
NP_057636.2
2
283
NM_017844.3
NP_060314.2
3
446
XM_011511300.2
XP_011509602.1
3
630
XM_011511301.1
XP_011509603.1
3
630
XM_011511304.2
XP_011509606.1
3
630
XM_011511305.2
XP_011509607.1
3
630
XM_011511309.2
XP_011509611.1
3
630
XM_011511313.2
XP_011509615.1
3
630
XM_011511315.2
XP_011509617.1
3
630
XM_011511317.1
XP_011509619.1
3
630
XM_011511318.1
XP_011509620.1
3
630
XM_011511319.2
XP_011509621.1
3
630
XM_011511322.1
XP_011509624.1
3
630
XM_017004271.1
XP_016859760.1
3
630
XM_017004272.1
XP_016859761.1
3
630
XM_017004273.1
XP_016859762.1
3
630
XM_017004274.1
XP_016859763.1
3
630
XM_017004275.1
XP_016859764.1
3
396
XM_017004276.1
XP_016859765.1
4
717
XM_017004277.1
XP_016859766.1
3
630
XM_017004278.1
XP_016859767.1
3
630
XM_017004279.1
XP_016859768.1
3
630
XM_017004280.1
XP_016859769.1
3
630
XM_017004281.1
XP_016859770.1
2
236
XM_017004282.1
XP_016859771.1
2
480
XM_017004283.1
XP_016859772.1
4
717
XM_017004284.1
XP_016859773.1
3
630
XM_017004285.1
XP_016859774.1
3
396
XM_017004286.1
XP_016859775.1
2
480
XM_017004287.1
XP_016859776.1
2
480
XM_017004288.1
XP_016859777.1
2
480
XM_017004289.1
XP_016859778.1
2
480
XM_017004290.1
XP_016859779.1
2
480
XM_017004291.1
XP_016859780.1
4
667
GenBank mRNA
AB034636.1
-
2
213
AK000506.1
-
3
413
AK125881.1
-
1
647
AK225534.1
-
3
413
AK296714.1
-
2
199
AK314534.1
-
3
430
AL136618.1
-
2
192
BC033495.1
-
3
446
BC073146.1
-
3
387
5743806
CR936600.1
-
2
181
CR936759.1
-
3
354
DB019614.1
-
3
685
KU178642.1
-
2
288
KU178643.1
-
2
288
Back To Top
More Information
Gene Ontology Categories:
Process(es)
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0