Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016563.3 | NP_057647.1 | 1, 2 | 383 | ||
XM_005254434.4 | XP_005254491.1 | 1, 2 | 193 | |||
XM_017022296.1 | XP_016877785.1 | 1, 2 | 193 | |||
GenBank mRNA | AF233588.1 | - | 1, 2 | 222 | ||
AK314914.1 | - | 1, 2 | 191 | |||
BC053734.1 | - | 1, 2 | 231 | 5588236 | ||
BI489884.1 | - | 1, 2 | 383 |