Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024021.3 | NP_076926.2 | 6 | 725 | ||
NM_148975.2 | NP_683876.1 | 5 | 579 | |||
XM_017017909.1 | XP_016873398.1 | 5 | 592 | |||
GenBank mRNA | AB013102.1 | - | 5 | 420 | ||
AB022821.1 | - | 5 | 444 | |||
AF068288.1 | - | 5 | 513 | |||
AF237912.1 | - | 5 | 530 | |||
AF350500.1 | - | 4 | 483 | |||
AK075081.1 | - | 6 | 610 | |||
AK292089.1 | - | 5 | 534 | |||
AK314033.1 | - | 5 | 422 | |||
BC020648.1 | - | 5 | 454 | 4720088 | ||
BG538851.1 | - | 5 | 581 |