Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UCHL5 SIRNA Assays ›
          Gene Symbol:
          UCHL5
          Gene Name:
          ubiquitin C-terminal hydrolase L5
          Gene Aliases:
          CGI-70, INO80R, UCH-L5, UCH37
          Chromosome Location:
          Chr.1: 193012366 - 193060140 on Build GRCh38
          UniGene ID:
          Hs.145469
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s28048
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UCHL5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199261.1 NP_001186190.1 9 902
          NM_001199262.1 NP_001186191.1 9 902
          NM_001199263.1 NP_001186192.1 9 902
          NM_015984.3 NP_057068.1 9 905
          NR_037607.1 9 1168
          XM_005245244.4 XP_005245301.1 10 1066
          XM_005245246.4 XP_005245303.1 9 985
          XM_005245247.4 XP_005245304.1 9 840
          XM_005245248.4 XP_005245305.1 9 860
          XM_005245249.4 XP_005245306.1 9 759
          XM_006711366.3 XP_006711429.1 10 1069
          XM_006711367.3 XP_006711430.1 10 1069
          XM_006711368.3 XP_006711431.1 10 1069
          XM_006711369.2 XP_006711432.1 10 1066
          XM_006711370.3 XP_006711433.3 10 1022
          XM_006711371.3 XP_006711434.1 10 840
          XM_011509604.1 XP_011507906.1 10 1069
          XM_011509607.2 XP_011507909.1 10 880
          XM_011509608.2 XP_011507910.1 11 1241
          XM_011509609.2 XP_011507911.1 11 1145
          XM_017001429.1 XP_016856918.1 10 1066
          XM_017001430.1 XP_016856919.1 10 1066
          XM_017001431.1 XP_016856920.1 9 988
          XM_017001432.1 XP_016856921.1 9 988
          XM_017001433.1 XP_016856922.1 9 988
          XM_017001434.1 XP_016856923.1 10 1019
          XM_017001435.1 XP_016856924.1 9 938
          XM_017001436.1 XP_016856925.1 10 1160
          XM_017001437.1 XP_016856926.1 10 1157
          XM_017001438.1 XP_016856927.1 10 864
          XM_017001439.1 XP_016856928.1 10 1282
          XM_017001440.1 XP_016856929.1 9 1168
          XM_017001441.1 XP_016856930.1 9 1141
          XM_017001442.1 XP_016856931.1 9 756
          XM_017001443.1 XP_016856932.1 9 759
          XM_017001444.1 XP_016856933.1 9 1150
          GenBank mRNA AF147717.1 - 9 903
          AF151828.1 - 9 782
          AF157320.1 - 9 917
          AK024751.1 - 9 848
          AK225794.1 - 9 838
          AK296487.1 - 5 492
          AK301381.1 - 9 1168
          AK314445.1 - 9 849
          AK316064.1 - 8 743
          BC015521.1 - 9 835 3912584
          BC025369.2 - 9 880 3937407
          BT006790.1 - 9 769

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00255374_m1

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          DNA recombination
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          negative regulation of endopeptidase activity
          protein deubiquitination
          lateral ventricle development
          midbrain development
          forebrain morphogenesis
          regulation of proteasomal protein catabolic process
          thiol-dependent ubiquitin-specific protease activity
          endopeptidase inhibitor activity
          protein binding
          poly(A) RNA binding
          proteasome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0