Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@4167e21d
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other UCHL5 SIRNA Assays ›
Gene Symbol:
UCHL5
Gene Name:
ubiquitin C-terminal hydrolase L5
Gene Aliases:
CGI-70, INO80R, UCH-L5, UCH37
Chromosome Location:
Chr.1: 193012366 - 193060140 on Build GRCh38
UniGene ID:
Hs.145469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s28049
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCHL5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199261.1
NP_001186190.1
8
774
NM_001199262.1
NP_001186191.1
8
774
NM_001199263.1
NP_001186192.1
8
774
NM_015984.3
NP_057068.1
8
774
NR_037607.1
8
1040
XM_005245244.4
XP_005245301.1
9
938
XM_005245246.4
XP_005245303.1
8
857
XM_005245247.4
XP_005245304.1
8
709
XM_005245248.4
XP_005245305.1
8
732
XM_005245249.4
XP_005245306.1
8
628
XM_006711366.3
XP_006711429.1
9
938
XM_006711367.3
XP_006711430.1
9
938
XM_006711368.3
XP_006711431.1
9
938
XM_006711369.2
XP_006711432.1
9
938
XM_006711370.3
XP_006711433.3
9
891
XM_006711371.3
XP_006711434.1
9
709
XM_011509604.1
XP_011507906.1
9
938
XM_011509607.2
XP_011507909.1
9
749
XM_011509608.2
XP_011507910.1
10
1110
XM_011509609.2
XP_011507911.1
10
1014
XM_017001429.1
XP_016856918.1
9
938
XM_017001430.1
XP_016856919.1
9
938
XM_017001431.1
XP_016856920.1
8
857
XM_017001432.1
XP_016856921.1
8
857
XM_017001433.1
XP_016856922.1
8
857
XM_017001434.1
XP_016856923.1
9
891
XM_017001435.1
XP_016856924.1
8
810
XM_017001436.1
XP_016856925.1
9
1029
XM_017001437.1
XP_016856926.1
9
1029
XM_017001438.1
XP_016856927.1
9
733
XM_017001439.1
XP_016856928.1
9
1154
XM_017001440.1
XP_016856929.1
8
1037
XM_017001441.1
XP_016856930.1
8
1013
XM_017001442.1
XP_016856931.1
8
628
XM_017001443.1
XP_016856932.1
8
628
XM_017001444.1
XP_016856933.1
8
1022
GenBank mRNA
AF147717.1
-
8
772
AF151828.1
-
8
654
AF157320.1
-
8
789
AK024751.1
-
8
720
AK225794.1
-
8
707
AK296487.1
-
4
364
AK301381.1
-
8
1040
AK314445.1
-
8
718
AK316064.1
-
7
612
BC015521.1
-
8
707
3912584
BC025369.2
-
8
752
3937407
BT006790.1
-
8
641
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00255374_m1
Panther Classification:
Molecular Function -
cysteine protease
hydrolase
protease
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-green:8080/10.253.229.154:8080.
git-commit: 0914912cb48844dc9118fffd0bcf8fc2c0fe1c71
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.17.0-2021.02.06-1.0