Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015986.3 | NP_057070.3 | 6 | 969 | ||
NR_073118.1 | 5 | 761 | ||||
XM_011524879.1 | XP_011523181.1 | 6 | 1143 | |||
GenBank mRNA | AF046059.1 | - | 6 | 882 | ||
AF120151.1 | - | 5 | 1335 | |||
AK296238.1 | - | 5 | 672 | |||
AK313315.1 | - | 6 | 958 | |||
BC023567.2 | - | 6 | 891 | 4309046 | ||
DQ298450.1 | - | 6 | 848 |