Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016224.4 | NP_057308.1 | 10 | 1325 | ||
XM_005267015.2 | XP_005267072.1 | 12 | 1403 | |||
XM_011535886.2 | XP_011534188.1 | 7 | 864 | |||
GenBank mRNA | AB209736.1 | - | 6 | 656 | ||
AF076957.1 | - | 5 | 545 | |||
AF121859.1 | - | 10 | 1085 | |||
AF131214.1 | - | 10 | 1104 | |||
AF172847.1 | - | 10 | 1205 | |||
AK026637.1 | - | 7 | 870 | |||
AK127345.1 | - | 7 | 1180 | |||
AK314360.1 | - | 10 | 1159 | |||
BC001084.2 | - | 1 | 126 | |||
BC005022.1 | - | 10 | 1232 | 3832234 |