Search
Search

RTCB| Gene Symbol: RTCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014306.4 | NP_055121.1 | 5 | 572 | ||
| GenBank mRNA | AF155658.1 | - | 5 | 520 | ||
| AF161466.1 | - | 5 | 531 | |||
| AK297826.1 | - | 5 | 449 | |||
| AK297908.1 | - | 532 | ||||
| AK312503.1 | - | 5 | 532 | |||
| AL050255.1 | - | 5 | 516 | |||
| BC000151.1 | - | 5 | 510 | 2984824 | ||
| BC002970.1 | - | 5 | 510 | 3533883 | ||
| BC010308.1 | - | 5 | 522 | 3451576 | ||
| BC016707.1 | - | 5 | 537 | 4102148 | ||
| CR456450.1 | - | 5 | 487 | |||
| CR457175.1 | - | 5 | 441 | |||