Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015934.4 | NP_057018.1 | 10 | 1164 | ||
GenBank mRNA | AF123534.1 | - | 10 | 938 | ||
AF263608.1 | - | 10 | 938 | |||
AK023975.1 | - | 9 | 863 | |||
AK307396.1 | - | 10 | 1144 | |||
AL117554.1 | - | 10 | 1147 | |||
BC001707.1 | - | 10 | 1130 | |||
BC009306.2 | - | 10 | 1094 | |||
BC018805.1 | - | 10 | 1117 | |||
BC032592.2 | - | 10 | 1088 | 5527148 |