Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERAP1 SIRNA Assays ›
Gene Symbol:
ERAP1
Gene Name:
endoplasmic reticulum aminopeptidase 1
Gene Aliases:
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
Chromosome Location:
Chr.5: 96759245 - 96935983 on Build GRCh38
UniGene ID:
Hs.666524
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s28616
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040458.1
NP_001035548.1
17
2874
NM_001198541.1
NP_001185470.1
17
2632
NM_016442.3
NP_057526.3
17
2874
XM_005272015.4
XP_005272072.1
17
2825
XM_005272016.4
XP_005272073.1
17
2785
XM_011543480.2
XP_011541782.1
21
5094
XM_011543481.2
XP_011541783.1
21
5091
XM_011543484.2
XP_011541786.1
21
5090
XM_011543485.2
XP_011541787.1
20
4910
XM_011543486.2
XP_011541788.1
21
5094
XM_017009580.1
XP_016865069.1
22
5150
XM_017009581.1
XP_016865070.1
20
4936
XM_017009582.1
XP_016865071.1
17
2609
XM_017009583.1
XP_016865072.1
17
2767
GenBank mRNA
AB011097.2
-
16
2548
AF106037.1
-
17
2589
AF183569.1
-
17
2594
AF222340.1
-
17
2614
AK222873.1
-
4
464
AK291636.1
-
17
2624
AK310535.1
-
16
2049
AY028806.1
-
17
2624
AY028807.1
-
17
2624
AY358590.1
-
17
2612
BC030775.1
-
17
2626
4824919
KM357870.1
-
16
2527
KM357871.1
-
16
2527
KM357872.1
-
16
2527
KM357873.1
-
16
2527
KM357874.1
-
16
2527
KM357875.1
-
16
2527
KM357876.1
-
16
2527
KM357877.1
-
16
2527
KM357878.1
-
16
2527
KM357879.1
-
16
2527
KM357880.1
-
16
2527
KM357881.1
-
16
2527
KM357882.1
-
16
2527
KM357883.1
-
16
2527
KM357884.1
-
16
2527
KM357885.1
-
16
2527
KM357886.1
-
16
2527
KM357887.1
-
16
2527
KM357888.1
-
16
2527
KM357889.1
-
16
2527
KM357890.1
-
16
2527
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00429970_m1
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
membrane protein ectodomain proteolysis
regulation of blood pressure
response to bacterium
antigen processing and presentation of endogenous peptide antigen via MHC class I
peptide catabolic process
regulation of innate immune response
fat cell differentiation
positive regulation of angiogenesis
endopeptidase activity
aminopeptidase activity
interleukin-6 receptor binding
interleukin-1, Type II receptor binding
tumor necrosis factor receptor binding
protein binding
metalloexopeptidase activity
zinc ion binding
peptide binding
metalloaminopeptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-flftk:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0