Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042452.1 | NP_001035917.1 | 7 | 806 | ||
NM_001042453.1 | NP_001035918.1 | 8 | 1052 | |||
NM_016542.3 | NP_057626.2 | 9 | 1283 | |||
XM_011531349.1 | XP_011529651.1 | 10 | 1256 | |||
XM_011531350.1 | XP_011529652.1 | 9 | 1329 | |||
GenBank mRNA | AB040057.2 | - | 9 | 1202 | ||
AF231012.1 | - | 9 | 1171 | |||
AF344882.1 | - | 8 | 992 | |||
AF344883.1 | - | 7 | 806 | |||
AK000556.1 | - | 5 | 615 | |||
AK027589.1 | - | 4 | 500 | |||
AK074837.1 | - | 4 | 536 | |||
AK075107.1 | - | 8 | 964 | |||
AK075150.1 | - | 9 | 1198 | |||
AK303583.1 | - | 10 | 1264 | |||
AK314356.1 | - | 9 | 1199 | |||
AK316137.1 | - | 10 | 1264 | |||
BC098315.1 | - | 9 | 1188 | 40026349 | ||
BC099843.1 | - | 9 | 1188 | 40026348 | ||
BC103503.1 | - | 9 | 1188 | 40026350 | ||
BT020099.1 | - | 8 | 982 |