Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM7SF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016551.2 | NP_057635.1 | 4 | 669 | ||
XM_005253391.3 | XP_005253448.1 | 4 | 690 | |||
XM_011520690.2 | XP_011518992.1 | 3 | 532 | |||
XM_011520691.1 | XP_011518993.1 | 4 | 690 | |||
XM_017019463.1 | XP_016874952.1 | 4 | 780 | |||
XM_017019464.1 | XP_016874953.1 | 2 | 342 | |||
XM_017019465.1 | XP_016874954.1 | 2 | 342 | |||
XM_017019466.1 | XP_016874955.1 | 3 | 509 | |||
GenBank mRNA | AB032470.1 | - | 4 | 468 | ||
AK002031.1 | - | 4 | 435 | |||
AK023085.1 | - | 4 | 657 | |||
BC005176.2 | - | 4 | 594 | 3505386 |