Search
Search

GALNT7| Gene Symbol: GALNT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017423.2 | NP_059119.2 | 9 | 1531 | ||
| XM_005263062.1 | XP_005263119.1 | 9 | 1531 | |||
| XM_011532025.2 | XP_011530327.1 | 9 | 2278 | |||
| XM_017008292.1 | XP_016863781.1 | 12 | 2451 | |||
| XM_017008293.1 | XP_016863782.1 | 12 | 4240 | |||
| GenBank mRNA | AJ002744.1 | - | 9 | 1480 | ||
| AK295509.1 | - | 7 | 837 | |||
| BC035303.1 | - | 8 | 962 | |||
| BC046129.1 | - | 9 | 1509 | 4442080 | ||
| BC047468.1 | - | 9 | 1506 | 5271749 | ||