Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286462.1 | NP_001273391.1 | 2 | 389 | ||
NM_001286463.1 | NP_001273392.1 | 1 | 200 | |||
NM_001286476.1 | NP_001273405.1 | 2 | 1462 | |||
NM_001286477.1 | NP_001273406.1 | 2 | 921 | |||
NM_058180.4 | NP_478060.2 | 2 | 1462 | |||
XM_005261148.4 | XP_005261205.1 | 2 | 1448 | |||
XM_006724018.3 | XP_006724081.1 | 2 | 1448 | |||
XM_011529619.2 | XP_011527921.1 | 2 | 1448 | |||
XM_011529620.2 | XP_011527922.1 | 2 | 1448 | |||
XM_011529623.2 | XP_011527925.1 | 2 | 3450 | |||
XM_011529625.2 | XP_011527927.1 | 2 | 599 | |||
XM_011529626.2 | XP_011527928.1 | 3 | 3627 | |||
XM_011529627.2 | XP_011527929.1 | 3 | 3627 | |||
XM_011529628.1 | XP_011527930.1 | 2 | 1448 | |||
XM_017028388.1 | XP_016883877.1 | 3 | 3627 | |||
XM_017028389.1 | XP_016883878.1 | 3 | 3627 | |||
XM_017028390.1 | XP_016883879.1 | 3 | 1625 | |||
XM_017028391.1 | XP_016883880.1 | 2 | 1448 | |||
XM_017028392.1 | XP_016883881.1 | 3 | 446 | |||
GenBank mRNA | AK056387.1 | - | 2 | 1435 | ||
AK096641.1 | - | 1 | 1117 | |||
AK098098.1 | - | 1 | 137 | |||
AY039243.1 | - | 1 | 186 | |||
AY039244.1 | - | 1 | 186 | |||
BC026042.1 | - | 2 | 1437 | |||
BC121007.1 | - | 3 | 1609 | |||
BC121008.1 | - | 2 | 1432 | 40116957 |