Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT4 SIRNA Assays ›
          Gene Symbol:
          WNT4
          Gene Name:
          Wnt family member 4
          Gene Aliases:
          SERKAL, WNT-4
          Chromosome Location:
          Chr.1: 22117305 - 22143981 on Build GRCh38
          UniGene ID:
          Hs.25766
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s28935

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030761.4 NP_110388.2 2 233
          XM_011541597.2 XP_011539899.1 2 1707
          XM_011541598.2 XP_011539900.1 2 302
          XM_011541599.1 XP_011539901.1 2 218
          GenBank mRNA AB061675.1 - 2 134
          AF316543.1 - 2 173
          AK296058.1 - 2 213
          AY009398.1 - 2 129
          AY358947.1 - 2 165
          BC057781.1 - 2 324 30330746
          BT020125.1 - 2 129

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_009523, M89797, AK012727.

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          branching involved in ureteric bud morphogenesis
          kidney development
          epithelial to mesenchymal transition
          embryonic epithelial tube formation
          liver development
          androgen biosynthetic process
          male gonad development
          female gonad development
          cellular response to starvation
          negative regulation of gene expression
          negative regulation of steroid biosynthetic process
          Wnt signaling pathway
          regulation of cell-cell adhesion
          neuron differentiation
          female sex determination
          adrenal gland development
          negative regulation of cell migration
          positive regulation of bone mineralization
          positive regulation of aldosterone biosynthetic process
          positive regulation of collagen biosynthetic process
          immature T cell proliferation in thymus
          non-canonical Wnt signaling pathway via MAPK cascade
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of GTPase activity
          cell fate commitment
          negative regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of meiotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          oocyte development
          smooth muscle cell differentiation
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          canonical Wnt signaling pathway
          somatotropin secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          mesenchymal to epithelial transition
          tertiary branching involved in mammary gland duct morphogenesis
          negative regulation of wound healing
          mammary gland epithelium development
          positive regulation of dermatome development
          paramesonephric duct development
          negative regulation of testicular blood vessel morphogenesis
          cellular response to transforming growth factor beta stimulus
          renal vesicle formation
          renal vesicle induction
          metanephric mesenchymal cell differentiation
          metanephric tubule formation
          metanephric nephron morphogenesis
          establishment of protein localization to plasma membrane
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          negative regulation of male gonad development
          positive regulation of cortisol biosynthetic process
          negative regulation of testosterone biosynthetic process
          negative regulation of apoptotic signaling pathway
          transcription corepressor activity
          frizzled binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0