Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRMT7 SIRNA Assays ›
Gene Symbol:
PRMT7
Gene Name:
protein arginine methyltransferase 7
Gene Aliases:
-
Chromosome Location:
Chr.16: 68310974 - 68358832 on Build GRCh38
UniGene ID:
Hs.640229
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29078
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRMT7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184824.1
NP_001171753.1
12
1400
NM_001290018.1
NP_001276947.1
14
2191
NM_019023.2
NP_061896.1
14
1685
XM_011523112.2
XP_011521414.1
13
1550
XM_011523113.2
XP_011521415.1
14
1677
XM_011523115.2
XP_011521417.1
13
1494
XM_011523116.2
XP_011521418.1
13
1550
XM_011523121.2
XP_011521423.1
11
1315
XM_011523124.2
XP_011521426.1
13
1603
XM_011523125.2
XP_011521427.1
12
1445
XM_011523126.2
XP_011521428.1
12
1449
XM_011523128.2
XP_011521430.1
13
1494
XM_011523131.2
XP_011521433.1
12
1424
XM_017023290.1
XP_016878779.1
14
1677
XM_017023291.1
XP_016878780.1
14
1617
XM_017023292.1
XP_016878781.1
13
1494
XM_017023293.1
XP_016878782.1
14
1677
XM_017023294.1
XP_016878783.1
14
1617
XM_017023295.1
XP_016878784.1
13
1550
XM_017023296.1
XP_016878785.1
14
1677
XM_017023297.1
XP_016878786.1
14
1677
XM_017023298.1
XP_016878787.1
12
1545
XM_017023299.1
XP_016878788.1
12
1545
XM_017023302.1
XP_016878791.1
12
1445
XM_017023303.1
XP_016878792.1
13
1603
XM_017023304.1
XP_016878793.1
14
1677
XM_017023305.1
XP_016878794.1
11
1315
XM_017023307.1
XP_016878796.1
13
1603
XM_017023308.1
XP_016878797.1
13
1617
XM_017023309.1
XP_016878798.1
12
1445
XM_017023310.1
XP_016878799.1
12
1469
XM_017023311.1
XP_016878800.1
12
1401
XM_017023313.1
XP_016878802.1
13
1592
XM_017023314.1
XP_016878803.1
12
1352
XM_017023315.1
XP_016878804.1
13
1592
XM_017023316.1
XP_016878805.1
12
1352
GenBank mRNA
AK001502.1
-
14
1617
AK022739.1
-
10
1112
AK056647.1
-
14
2173
AK097175.1
-
12
2127
AK304605.1
-
12
1323
BC000146.1
-
12
1426
2900965
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal snRNP assembly
regulation of gene expression by genetic imprinting
transcription, DNA-templated
regulation of transcription, DNA-templated
histone methylation
peptidyl-arginine methylation
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
cell differentiation
histone arginine methylation
DNA methylation involved in gamete generation
regulation of protein binding
histone H4-R3 methylation
histone-arginine N-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
protein-arginine N-methyltransferase activity
[myelin basic protein]-arginine N-methyltransferase activity
protein-arginine omega-N monomethyltransferase activity
protein-arginine omega-N asymmetric methyltransferase activity
protein-arginine omega-N symmetric methyltransferase activity
histone binding
ribonucleoprotein complex binding
histone methyltransferase activity (H4-R3 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0