Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UGT1A10 SIRNA Assays ›
Gene Symbol:
UGT1A10
UGT1A7
UGT1A8
Gene Name:
UDP glucuronosyltransferase family 1 member A10
UDP glucuronosyltransferase family 1 member A7
UDP glucuronosyltransferase family 1 member A8
Gene Aliases:
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
Chromosome Location:
Chr.2: 233636477 - 233773305 on Build GRCh38
Chromosome Location:
Chr.2: 233681938 - 233773299 on Build GRCh38
Chromosome Location:
Chr.2: 233617645 - 233773299 on Build GRCh38
UniGene ID:
Hs.554822
Hs.554822
Hs.554822
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID
s29218
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UGT1A10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019075.2
NP_061948.1
1
442
GenBank mRNA
AK025403.1
-
1
49
AY435137.1
-
1
396
BC020971.1
-
1
465
3850654
BC053576.1
-
1
417
4452440
BC069210.1
-
1
426
4660540
U89508.1
-
1
396
Gene Symbol: UGT1A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019077.2
NP_061950.2
1
396
GenBank mRNA
AY435142.1
-
1
396
U89507.1
-
1
396
Gene Symbol: UGT1A8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019076.4
NP_061949.3
1
459
GenBank mRNA
AF462267.1
-
1
429
AF462268.1
-
1
459
AK313488.1
-
1
442
AY435143.1
-
1
396
DQ364251.1
-
1
51
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
flavonoid biosynthetic process
flavone metabolic process
cellular glucuronidation
flavonoid glucuronidation
xenobiotic glucuronidation
excretion
coumarin metabolic process
drug metabolic process
retinoic acid metabolic process
negative regulation of catalytic activity
fatty acid metabolic process
steroid metabolic process
negative regulation of fatty acid metabolic process
negative regulation of steroid metabolic process
retinoic acid binding
protein kinase C binding
glucuronosyltransferase activity
protein homodimerization activity
protein heterodimerization activity
enzyme inhibitor activity
steroid binding
fatty acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System