Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UGT1A8 SIRNA Assays ›
          Gene Symbol:
          UGT1A8 UGT1A10 UGT1A7 UGT1A9
          Gene Name:
          UDP glucuronosyltransferase family 1 member A8
          UDP glucuronosyltransferase family 1 member A10
          UDP glucuronosyltransferase family 1 member A7
          UDP glucuronosyltransferase family 1 member A9
          Gene Aliases:
          UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
          UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
          UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
          HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I
          Chromosome Location:
          Chr.2: 233617645 - 233773299 on Build GRCh38
          Chromosome Location:
          Chr.2: 233636477 - 233773305 on Build GRCh38
          Chromosome Location:
          Chr.2: 233681938 - 233773299 on Build GRCh38
          Chromosome Location:
          Chr.2: 233671898 - 233773305 on Build GRCh38
          UniGene ID:
          Hs.554822
          Hs.554822
          Hs.554822
          Hs.554822
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s29221

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UGT1A8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019076.4 NP_061949.3 1 816
          GenBank mRNA AF030310.1 - 1 753
          AF462267.1 - 1 786
          AF462268.1 - 1 816
          AK313488.1 - 1 799
          AY435143.1 - 1 753
          DQ364251.1 - 1 408
          Gene Symbol: UGT1A10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019075.2 NP_061948.1 1 799
          GenBank mRNA AK025403.1 - 1 406
          AY435137.1 - 1 753
          BC020971.1 - 1 822 3850654
          BC053576.1 - 1 774 4452440
          BC069210.1 - 1 783 4660540
          U89508.1 - 1 753
          Gene Symbol: UGT1A7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019077.2 NP_061950.2 1 753
          GenBank mRNA AY435142.1 - 1 753
          BC001521.2 - 1 323
          DQ383513.1 - 1 465
          U89507.1 - 1 753
          Gene Symbol: UGT1A9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021027.2 NP_066307.1 1 790
          GenBank mRNA AF056188.1 - 1 771
          AK313510.1 - 1 829
          AY435144.1 - 1 753
          BC058844.1 - 1 796 4593613
          DQ364246.1 - 1 105
          S55985.1 - 1 787

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid metabolic process
          steroid metabolic process
          coumarin metabolic process
          retinoic acid metabolic process
          negative regulation of catalytic activity
          negative regulation of fatty acid metabolic process
          negative regulation of steroid metabolic process
          flavone metabolic process
          flavonoid glucuronidation
          xenobiotic glucuronidation
          flavonoid biosynthetic process
          cellular glucuronidation
          excretion
          drug metabolic process
          xenobiotic metabolic process
          metabolic process
          retinoic acid binding
          enzyme inhibitor activity
          steroid binding
          fatty acid binding
          glucuronosyltransferase activity
          protein homodimerization activity
          protein heterodimerization activity
          protein kinase C binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zkdp8:80/100.66.75.244:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0