Search
Search

CETN2| Gene Symbol: CETN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004344.1 | NP_004335.1 | 2 | 108 | ||
| GenBank mRNA | AK311940.1 | - | 2 | 103 | ||
| BC005334.1 | - | 2 | 108 | 3961448 | ||
| BC013873.2 | - | 2 | 106 | 3836808 | ||
| BT007256.1 | - | 1 | 61 | |||
| X72964.1 | - | 2 | 108 | |||