Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DYM SIRNA Assays ›
Gene Symbol:
DYM
Gene Name:
dymeclin
Gene Aliases:
DMC, SMC
Chromosome Location:
Chr.18: 49041474 - 49460709 on Build GRCh38
UniGene ID:
Hs.162996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29481
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DYM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017653.3
NP_060123.3
14
1896
XM_006722488.2
XP_006722551.1
14
1891
XM_006722490.2
XP_006722553.1
14
1891
XM_011526036.1
XP_011524338.1
14
1891
XM_011526037.1
XP_011524339.1
14
1893
XM_011526038.1
XP_011524340.1
14
1888
XM_011526039.1
XP_011524341.1
14
1891
XM_011526040.1
XP_011524342.1
15
1892
XM_011526041.1
XP_011524343.1
14
1891
XM_011526042.1
XP_011524344.1
14
1891
XM_017025795.1
XP_016881284.1
14
1890
XM_017025796.1
XP_016881285.1
15
1902
XM_017025797.1
XP_016881286.1
14
1829
XM_017025798.1
XP_016881287.1
14
1888
XM_017025799.1
XP_016881288.1
14
1891
XM_017025800.1
XP_016881289.1
14
1888
XM_017025801.1
XP_016881290.1
14
1890
GenBank mRNA
AK000078.1
-
10
1209
AK074611.1
-
14
1891
AK091256.1
-
15
1865
AK225205.1
-
10
1209
AK291303.1
-
14
1861
AK296579.1
-
9
1283
AK315091.1
-
15
1837
AL390156.1
-
3
198
AY364250.1
-
13
1531
BC001252.1
-
11
1295
BC064394.1
-
14
1878
5752466
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
bone development
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0