Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193307.1 | NP_001180236.1 | 2 | 1220 | ||
NM_017654.3 | NP_060124.2 | 3 | 1321 | |||
GenBank mRNA | AB095925.2 | - | 3 | 1255 | ||
AF445355.1 | - | 3 | 1321 | |||
AK122951.1 | - | 2 | 1158 | |||
BC065769.1 | - | 3 | 1311 | |||
BC132773.1 | - | 2 | 1151 | 40146795 | ||
BC132775.1 | - | 2 | 1151 | 40146797 | ||
BC150249.1 | - | 3 | 1276 | 9007474 | ||
BX647072.1 | - | 3 | 1277 |