Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCAS3 SIRNA Assays ›
          Gene Symbol:
          BCAS3
          Gene Name:
          BCAS3, microtubule associated cell migration factor
          Gene Aliases:
          GAOB1, MAAB
          Chromosome Location:
          Chr.17: 60677811 - 61392838 on Build GRCh38
          UniGene ID:
          Hs.655028
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29533

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCAS3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099432.1 NP_001092902.1 8, 9 701
          NM_001320470.1 NP_001307399.1 8, 9 701
          NM_017679.3 NP_060149.3 8, 9 701
          XM_005257472.1 XP_005257529.1 8, 9 652
          XM_011524939.1 XP_011523241.1 8, 9 652
          XM_011524940.1 XP_011523242.1 8, 9 652
          XM_011524941.1 XP_011523243.1 8, 9 652
          XM_011524942.2 XP_011523244.1 8, 9 652
          XM_011524943.2 XP_011523245.1 8, 9 652
          XM_011524944.1 XP_011523246.1 8, 9 652
          XM_011524945.2 XP_011523247.1 8, 9 652
          XM_011524946.2 XP_011523248.1 8, 9 652
          XM_011524947.1 XP_011523249.1 8, 9 652
          XM_011524948.2 XP_011523250.1 8, 9 652
          XM_011524949.2 XP_011523251.1 8, 9 652
          XM_011524950.1 XP_011523252.1 8, 9 652
          XM_011524951.2 XP_011523253.1 8, 9 652
          XM_017024783.1 XP_016880272.1 8, 9 652
          XM_017024784.1 XP_016880273.1 8, 9 652
          XM_017024785.1 XP_016880274.1 8, 9 652
          XM_017024786.1 XP_016880275.1 8, 9 652
          XM_017024787.1 XP_016880276.1 8, 9 652
          XM_017024788.1 XP_016880277.1 8, 9 652
          XM_017024789.1 XP_016880278.1 8, 9 652
          XM_017024790.1 XP_016880279.1 8, 9 652
          XM_017024791.1 XP_016880280.1 8, 9 652
          XM_017024792.1 XP_016880281.1 8, 9 652
          XM_017024793.1 XP_016880282.1 8, 9 652
          XM_017024794.1 XP_016880283.1 8, 9 652
          XM_017024795.1 XP_016880284.1 8, 9 652
          GenBank mRNA AF361219.1 - 9, 10 640
          AK225757.1 - 8, 9 660
          AK314744.1 - 8, 9 646
          BC017390.1 - 8, 9 657
          BC117275.1 - 8, 9 623 40125826
          BC143386.1 - 8, 9 623 9051892
          BC143387.1 - 8, 9 623 9051893

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          transcription, DNA-templated
          Golgi organization
          positive regulation of endothelial cell migration
          vesicle-mediated transport
          microtubule organizing center organization
          negative regulation of GTPase activity
          tube formation
          response to starvation
          positive regulation of catalytic activity
          positive regulation of GTPase activity
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of filopodium assembly
          negative regulation of focal adhesion assembly
          cellular response to estrogen stimulus
          positive regulation of intracellular protein transport
          activation of GTPase activity
          regulation of establishment of cell polarity
          positive regulation of actin cytoskeleton reorganization
          chromatin binding
          protein binding
          transcription factor binding
          acetyltransferase activator activity
          histone acetyltransferase binding
          nuclear hormone receptor binding
          histone binding
          beta-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0