Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APTX SIRNA Assays ›
          Gene Symbol:
          APTX
          Gene Name:
          aprataxin
          Gene Aliases:
          AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
          Chromosome Location:
          Chr.9: 32883872 - 33025129 on Build GRCh38
          UniGene ID:
          Hs.20158
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29558

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APTX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195248.1 NP_001182177.1 7 941
          NM_001195249.1 NP_001182178.1 7 1110
          NM_001195250.1 NP_001182179.1 6 779
          NM_001195251.1 NP_001182180.1 8 1047
          NM_001195252.1 NP_001182181.1 7 725
          NM_001195254.1 NP_001182183.1 6 801
          NM_175069.2 NP_778239.1 7 941
          NM_175073.2 NP_778243.1 8 1047
          NR_036576.1 7 936
          NR_036577.1 7 826
          NR_036578.1 7 958
          NR_036579.1 7 1105
          XM_006716791.3 XP_006716854.1 7 949
          XM_006716792.2 XP_006716855.1 7 967
          XM_011517938.1 XP_011516240.1 6 925
          XM_011517939.2 XP_011516241.1 6 880
          XM_017014837.1 XP_016870326.1 6 1030
          XM_017014838.1 XP_016870327.1 7 923
          GenBank mRNA AB603741.1 - 6 837
          AK000164.1 - 6 944
          AK055672.1 - 7 928
          AK131046.1 - 7 893
          AY040777.1 - 7 916
          AY208829.1 - 7 916
          AY208830.1 - 6 779
          AY208831.1 - 7 691
          AY208832.1 - 6 754
          AY208833.1 - 6 754
          AY208834.1 - 6 855
          AY208835.1 - 7 896
          AY208836.1 - 7 1056
          AY208837.1 - 8 1013
          AY208838.1 - 6 872
          AY208839.1 - 7 1050
          AY208840.1 - 7 913
          AY208841.1 - 6 750
          AY208842.1 - 4 550
          AY302067.1 - 6 1059
          AY302068.1 - 6 1054
          AY302069.1 - 6 944
          AY302070.1 - 6 939
          AY302071.1 - 5 832
          AY302072.1 - 5 827
          AY302073.1 - 5 717
          AY302074.1 - 5 712
          BC001628.1 - 6 764
          BC032650.1 - 7 982 5505120
          BC068107.1 - 8 1022
          BC104881.1 - 7 955 8143884
          BX538161.1 - 8 1026

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          damaged DNA-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          DNA ligation
          cellular response to DNA damage stimulus
          dephosphorylation
          regulation of protein stability
          response to hydrogen peroxide
          nucleic acid phosphodiester bond hydrolysis
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          double-stranded RNA binding
          protein binding
          phosphoglycolate phosphatase activity
          DNA 5'-adenosine monophosphate hydrolase activity
          metal ion binding
          protein N-terminus binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0