Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017811.3 | NP_060281.2 | 4, 5 | 1064 | ||
XM_005251496.2 | XP_005251553.1 | 3, 4 | 997 | |||
GenBank mRNA | AJ240087.1 | - | 4, 5 | 495 | ||
AK000426.1 | - | 4, 5 | 681 | |||
AK225880.1 | - | 4, 5 | 669 | |||
BC004862.2 | - | 4, 5 | 671 | 3838157 | ||
BC047584.1 | - | 4, 5 | 920 | 4827644 | ||
CR457233.1 | - | 4, 5 | 490 |