Search
Search

MARC2| Gene Symbol: MARC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317338.1 | NP_001304267.1 | 5 | 1073 | ||
| NM_017898.4 | NP_060368.2 | 5 | 1073 | |||
| XM_006711407.2 | XP_006711470.1 | 4 | 1629 | |||
| XM_011509684.1 | XP_011507986.1 | 5 | 597 | |||
| GenBank mRNA | AK000612.1 | - | 3 | 212 | ||
| BC010366.1 | - | 4 | 407 | |||
| BC011973.1 | - | 5 | 985 | 3458649 | ||
| BC015829.2 | - | 3 | 343 | |||
| BC016859.1 | - | 5 | 975 | 3849257 | ||